missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzeneboronic acid, 97%

Product Code. 11495437
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495437 1 g 1g
11405447 5 g 5g
11415447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495437 Supplier Thermo Scientific Alfa Aesar Supplier No. B23454.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98546-51-1 | C7H9BO2S | 168.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 98546-51-1
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.017
MDL Number MFCD00093410
InChI Key IVUHTLFKBDDICS-UHFFFAOYSA-N
Synonym 4-methylthio phenylboronic acid, 4-thioanisoleboronic acid, 4-methylsulfanyl phenylboronic acid, 4-methylthiophenylboronic acid, 4-methylthio phenyl boronic acid, 4-methylsulfanylphenyl boronic acid, 4-methylthio phenylboronicacid, 4-methylthio benzeneboronic acid, thioanisole-4-boronic acid, 4-methylmercapto benzeneboronic acid
PubChem CID 2733959
IUPAC Name (4-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)SC)(O)O

Tekniske data

Melting Point ∼208°C to 212°C
Quantity 1 g
Beilstein 3247219
Formula Weight 168.02
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzeneboronic acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.