missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylthio)benzaldehyde, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 4-(Methylthio)benzaldehyde |
| CAS | 3446-89-7 |
| Molecular Formula | C8H8OS |
| Quantity | 100 g |
| Synonym | 4-(Methylmercapto)benzaldehyde, 4-Methanesulfanylbenzaldehyde, 4-Methanethiobenzaldehyde, 4-Methylsulfanylbenzaldehyde, Methyl 4-Formylphenyl thioether, p-(Methylthio)benzaldehyde, p-Methylthiobenzaldehyde |
| InChI Formula | InChI=1S/C8H8OS/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3 |
| SMILES | CSc1ccc(C=O)cc1 |
| IUPAC Name | 4-methylsulfanylbenzaldehyde |
| Molecular Weight (g/mol) | 152.21 |
| Formula Weight | 152.0296 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction