missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylthio)acetophenone, 99%

Product Code. 11334905
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334905 5 g 5g
11344905 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334905 Supplier Thermo Scientific Alfa Aesar Supplier No. L18074.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1778-09-2 | C9H10OS | 166.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1778-09-2
Molecular Formula C9H10OS
Molecular Weight (g/mol) 166.24
MDL Number MFCD00039835
InChI Key JECUZQLBQKNEMW-UHFFFAOYSA-N
Synonym 4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone
PubChem CID 74501
IUPAC Name 1-(4-methylsulfanylphenyl)ethanone
SMILES CSC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 80°C to 82°C
Boiling Point 135°C (1 mmHg)
Quantity 5 g
Beilstein 1860517
Formula Weight 166.24
Percent Purity 99%
Chemical Name or Material 4'-(Methylthio)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.