missing translation for 'onlineSavingsMsg'
Learn More

4'-(Methylthio)acetophenone 97%, ACROS Organics™

Product Code. 10065553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10065553 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065553 Supplier Thermo Scientific Acros Supplier No. 392650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1778-09-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 80°C to 84°C
Color Pink to Yellow or White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H10OS
Linear Formula CH3SC6H4C(=O)CH3
MDL Number MFCD00039835
Quantity 5 g
Synonym 4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone
InChI Key JECUZQLBQKNEMW-UHFFFAOYSA-N
SMILES CSC1=CC=C(C=C1)C(C)=O
IUPAC Name 1-[4-(methylsulfanyl)phenyl]ethan-1-one
Molecular Weight (g/mol) 166.24
PubChem CID 74501
Formula Weight 166.24
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4'-(Methylthio)acetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.