missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine, 97%, Thermo Scientific™

Catalog Number 10203422
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175204-61-2
C8H8N4S2
224.3
MFCD00052772
DKBHZJZOKUMDEF-UHFFFAOYSA-N
4-methylthio-6-2-thienyl-1,3,5-triazin-2-amine, 4-methylsulfanyl-6-thiophen-2-yl-1,3,5-triazin-2-amine, 4-methlythio-6-2-thienyl-1,3,5-triazin-2-amine, 1,3,5-triazin-2-amine,4-methylthio-6-2-thienyl, maybridge1_006475, 4-methylthio-6-2-thienyl-1,3,5-triazine-2-ylamine, 4-methylthio-6-thiophen-2-yl-1,3,5-triazin-2-amine
2782092
4-methylsulfanyl-6-thiophen-2-yl-1,3,5-triazin-2-amine
CSC1=NC(=NC(=N1)N)C2=CC=CS2
This item is not returnable. View return policy
175204-61-2
C8H8N4S2
224.3
MFCD00052772
DKBHZJZOKUMDEF-UHFFFAOYSA-N
4-methylthio-6-2-thienyl-1,3,5-triazin-2-amine, 4-methylsulfanyl-6-thiophen-2-yl-1,3,5-triazin-2-amine, 4-methlythio-6-2-thienyl-1,3,5-triazin-2-amine, 1,3,5-triazin-2-amine,4-methylthio-6-2-thienyl, maybridge1_006475, 4-methylthio-6-2-thienyl-1,3,5-triazine-2-ylamine, 4-methylthio-6-thiophen-2-yl-1,3,5-triazin-2-amine
2782092
4-methylsulfanyl-6-thiophen-2-yl-1,3,5-triazin-2-amine
CSC1=NC(=NC(=N1)N)C2=CC=CS2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 224.31
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)-6-(2-thienyl)-1, 3, 5-triazin-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'