missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15455407
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455407 1 g 1g
15465407 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455407 Supplier Thermo Scientific Alfa Aesar Supplier No. H32662.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 20485-41-0
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD00626872
InChI Key ZGWGSEUMABQEMD-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
PubChem CID 209805
IUPAC Name 4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC=N1)C(=O)O

Specifica

Melting Point 287°C
Quantity 1 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.