missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3586785
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
20485-41-0
C5H5NO2S
143.16
MFCD00626872
ZGWGSEUMABQEMD-UHFFFAOYSA-N
4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
209805
4-methyl-1,3-thiazole-5-carboxylic acid
CC1=C(SC=N1)C(=O)O
This item is not returnable. View return policy
20485-41-0
C5H5NO2S
143.16
MFCD00626872
ZGWGSEUMABQEMD-UHFFFAOYSA-N
4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
209805
4-methyl-1,3-thiazole-5-carboxylic acid
CC1=C(SC=N1)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'