missing translation for 'onlineSavingsMsg'
Learn More

4-Methyltetrahydro-1(2H)-pyrazinecarboximidamide hydroiodide, 97%, Thermo Scientific™

Product Code. 11386885
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 77723-03-6
Molecular Formula C6H15IN4
Molecular Weight (g/mol) 270.118
MDL Number MFCD09025920
InChI Key KESCAEZAONDSRV-UHFFFAOYSA-N
Synonym 4-methylpiperazine-1-carboximidamide hydroiodide, 4-methyltetrahydro-1 2h-pyrazinecarboximidamide hydroiodide, 4-methylpiperazinecarboxamidine, iodide, 4-methyl-1-piperazinecarboximidamide hydroiodide, 4-methylpiperazine-1-carboximidamide-hydrogen iodide 1/1
PubChem CID 20284359
IUPAC Name 4-methylpiperazine-1-carboximidamide;hydroiodide
SMILES CN1CCN(CC1)C(=N)N.I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 270.11
Percent Purity 97%
Chemical Name or Material 4-Methyltetrahydro-1(2H)-pyrazinecarboximidamide hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'