missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonylbenzylamine hydrochloride, 95%

Product Code. 10730882 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10730882 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10730882 Supplier Thermo Scientific Acros Supplier No. 424950025
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Formats
Denna artikel kan inte returneras. Se returpolicy

CAS: 4393-16-2 | C8H11NO2S | 185.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 4393-16-2
CAS Min % 94.0
CAS Max % 100.0
Melting Point 269°C to 277°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C8H11NO2S
Linear Formula H2NH2CC6H4SO2CH3·HCl
MDL Number MFCD00216491
Quantity 2.5 g
Synonym 4-methylsulfonyl benzylamine, 4-methanesulfonylphenyl methanamine, 4-methylsulfonyl phenyl methanamine, 4-methylsulfonylbenzylamine, benzenemethanamine, 4-methylsulfonyl, unii-61o466wzw2, 4-methanesulfonyl benzylamine, 4-methylsulfonyl benzenemethanamine, 4-methanesulfonylphenyl methylamine, 4-methanesulfonyl-benzylamine
InChI Key VMNXLLDFGVEBLE-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)CN
IUPAC Name (4-methylsulfonylphenyl)methanamine
Molecular Weight (g/mol) 185.241
PubChem CID 57421
Formula Weight 221.71
Percent Purity ≥94% (Acid)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylsulphonylbenzylamine hydrochloride, 95%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.