missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonylbenzyl bromide, Tech.

Product Code. 10267553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10267553 10 g Glass bottle 10g
10632002 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10267553 Supplier Thermo Scientific Acros Supplier No. 357580100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53606-06-7 | C8H9BrO2S | 249.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53606-06-7
Molecular Formula C8H9BrO2S
Molecular Weight (g/mol) 249.13
MDL Number MFCD00185849
InChI Key HGKPAXHJTMHWAH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
PubChem CID 2733581
IUPAC Name 1-(bromomethyl)-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)CBr

Specifications

Melting Point 82°C to 86°C
Color Beige to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 06, IV, 2210
Formula Weight 249.13
Percent Purity ≥60%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylsulphonylbenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylsulphonylbenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.