missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonylaniline hydrochloride, 95%, Thermo Scientific™

Product Code. 10637801 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 10637801

Brand: Thermo Scientific Acros 424930025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 177662-76-9
CAS Min % 94.0
CAS Max % 100.0
Melting Point 243.0°C to 245.0°C
Color Gray-Purple or White
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C7H9NO2S
Linear Formula H2NC6H4SO2CH3·HCl
MDL Number MFCD00216490
Quantity 2.5 g
Synonym 4-methylsulfonylaniline hydrochloride, 4-methylsulfonyl aniline hydrochloride, 4-methanesulfonylaniline hydrochloride, 4-methylsulphonylaniline hydrochloride, 4-methylsulphonyl aniline hydrochloride, 4-methanesulfonyl aniline hydrochloride, 4 methanesulfonyl aniline hydrochloride, acmc-1c3bo, 4-methanesulonyl aniline hydrochloride, 4-methanesulfonylphenylamine hydrochloride
Solubility Information Solubility in water: slight
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(N)C=C1
IUPAC Name 4-methanesulfonylaniline
Molecular Weight (g/mol) 171.21
PubChem CID 2735180
Formula Weight 207.68
Percent Purity 95%
Physical Form Crystals or Needles
Chemical Name or Material 4-Methylsulphonylaniline hydrochloride, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.