missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonylacetophenone, 95%, ACROS Organics™

Product Code. 10279900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
2.5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10279900 2.5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10279900 Supplier Thermo Scientific Acros Supplier No. 424910025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10297-73-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 126°C to 131°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C9H10O3S
Linear Formula H3CC(O)C6H4SO2CH3
MDL Number MFCD00025072
Quantity 2.5 g
Synonym 4'-methylsulfonyl acetophenone, 1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonylacetophenone, 4-methylsulfonyl acetophenone, 4-acetylphenyl methyl sulphone, 1-4-methylsulfonylphenyl ethanone, 4-methylsulphonylacetophenone, 1-4-methanesulfonyl-phenyl-ethanone, p-methylsulfonylacetophenone, 4-methanesulfonylacetophenone
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
Molecular Weight (g/mol) 198.236
PubChem CID 82529
Formula Weight 198.24
Percent Purity ≥94%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Methylsulphonylacetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.