missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonylamino)benzonitrile, 97%, Thermo Scientific™

Product Code. 11444397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444397 1 g 1g
11454397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444397 Supplier Thermo Scientific Alfa Aesar Supplier No. B20439.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36268-67-4
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD00204724
InChI Key BRDHOCVMWSXEHI-UHFFFAOYSA-N
Synonym 4-methanesulfonylamino benzonitrile, n-4-cyanophenyl methanesulfonamide, 4-methylsulfonylamino benzonitrile, methanesulfonamide, n-4-cyanophenyl, 4-methylsulfonyl amino benzenecarbonitrile, acmc-20amud, 4-methylsulfonyl amino benzonitrile, methanesulfonamide,n-4-cyanophenyl
PubChem CID 2734199
IUPAC Name N-(4-cyanophenyl)methanesulfonamide
SMILES CS(=O)(=O)NC1=CC=C(C=C1)C#N

Specifications

Melting Point 192°C to 196°C
Quantity 1 g
UN Number UN3439
Formula Weight 196.23
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonylamino)benzonitrile
compliance-icons
Product Identifier
  • 4-(Methanesulfonylamino)benzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Methanesulfonylamino)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.