missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzoic acid, 98%

Product Code. 15437867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437867 5 g 5g
15427867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437867 Supplier Thermo Scientific Alfa Aesar Supplier No. H50230.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4052-30-6 | C8H8O4S | 200.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4052-30-6
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00007564
InChI Key AJBWNNKDUMXZLM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzoic acid, p-methylsulfonylbenzoic acid, 4-methanesulfonyl-benzoic acid, 4-methylsulphonylbenzoic acid, 4-methylsulphonyl benzoic acid, 4-methanesulfonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulphonyl benzoic acid, 4-methanesulfonyl benzoic acid
PubChem CID 77670
IUPAC Name 4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 270°C to 275°C
Quantity 5 g
Formula Weight 200.21
Percent Purity 98%
Chemical Name or Material 4-(Methylsulfonyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.