missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)aniline, TRC

Product Code. 30473968
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473968 10 g 10g
30484143 5 g 5g
30482674 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473968 Supplier Toronto Research Chemicals Supplier No. TRCM33980810G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-49-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.22
IUPAC Name 4-methylsulfonylaniline
SMILES CS(=O)(=O)c1ccc(N)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H9NO2S/c1-11(9,10)7-4-2-6(8)3-5-7/h2-5H,8H2,1H3
Formula Weight 171.0354
Chemical Name or Material 4-(Methylsulfonyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.