missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)aniline hydrochloride, 99%

Product Code. p-7026899
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11470797

Brand: Thermo Scientific Alfa Aesar B23202.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 177662-76-9 | C7H9NO2S | 171.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177662-76-9
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00216490
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
Synonym 4-methylsulfonylaniline hydrochloride, 4-methylsulfonyl aniline hydrochloride, 4-methanesulfonylaniline hydrochloride, 4-methylsulphonylaniline hydrochloride, 4-methylsulphonyl aniline hydrochloride, 4-methanesulfonyl aniline hydrochloride, 4 methanesulfonyl aniline hydrochloride, acmc-1c3bo, 4-methanesulonyl aniline hydrochloride, 4-methanesulfonylphenylamine hydrochloride
PubChem CID 2735180
IUPAC Name 4-methanesulfonylaniline
SMILES CS(=O)(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 237°C to 239°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 207.67
Percent Purity 99%
Chemical Name or Material 4-(Methylsulfonyl)aniline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.