missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulfamoylbenzeneboronic acid, 97%

Product Code. 15499237
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499237 250 mg 250mg
15489237 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499237 Supplier Thermo Scientific Alfa Aesar Supplier No. H52493.MD
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 226396-31-2 | C7H10BNO4S | 215.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226396-31-2
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD06659864
InChI Key DOQOQZHSIBBHMY-UHFFFAOYSA-N
Synonym 4-n-methylsulfamoyl phenyl boronic acid, methyl 4-boronobenzenesulfonamide, 4-methylsulfamoyl phenyl boronic acid, 4-methylaminosulphonyl benzene boronic acid, 4-n-methylsulfamoyl phenylboronic acid, 4-methylsulfamoyl phenylboronic acid, 4-methylsulfamoylphenylboronic acid, boronic acid, 4-methylamino sulfonyl phenyl, acmc-209fxs
PubChem CID 44118763
IUPAC Name [4-(methylsulfamoyl)phenyl]boronic acid
SMILES CNS(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 136°C to 140°C
Quantity 250 mg
Formula Weight 215.03
Percent Purity 97%
Chemical Name or Material 4-Methylsulfamoylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.