missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsalicylic acid, 99%

Product Code. 11352116
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352116 25 g 25g
11362116 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352116 Supplier Thermo Scientific Alfa Aesar Supplier No. L03367.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-85-1 | C8H8O3 | 152.149 g/mol

4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

Solubility
Soluble in water 10 g/L @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-85-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002454
InChI Key NJESAXZANHETJV-UHFFFAOYSA-N
Synonym 4-methylsalicylic acid, m-cresotic acid, 2,4-cresotic acid, m-cresotinic acid, 2-hydroxy-p-toluic acid, m-homosalicylic acid, 4-methyl-2-hydroxybenzoic acid, gamma-cresotic acid, benzoic acid, 2-hydroxy-4-methyl, 4-methylsalicylate
PubChem CID 5788
ChEBI CHEBI:20450
IUPAC Name 2-hydroxy-4-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)C(=O)O)O

Specifications

Melting Point 176°C to 180°C
Odor Odorless
Quantity 25 g
Beilstein 2208140
Merck Index 14,2580
Solubility Information Soluble in water 10g/L @ 20?C.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 4-Methylsalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.