missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylquinazolin-2-yl)methanol, TRC

Product Code. 30468251
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
30468251 100 mg 100mg
30478189 50 mg 50mg
30465438 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30468251

Brand: Toronto Research Chemicals TRCM329338100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13535-91-6
Formula Weight 174.0793
InChI Formula InChI=1S/C10H10N2O/c1-7-8-4-2-3-5-9(8)12-10(6-13)11-7/h2-5,13H,6H2,1H3
IUPAC Name (4-methylquinazolin-2-yl)methanol
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.2
SMILES Cc1nc(CO)nc2ccccc12
Synonym 2-Hydroxymethyl-4-methylquinazoline
Chemical Name or Material (4-Methylquinazolin-2-yl)methanol

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.