missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyridin-2-amine, Mikromol™

Product Code. 30461228
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30461228

Brand: Mikromol MM3389.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methylpyridin-2-amine, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylpyridin-2-amine
CAS 695-34-1
Analyte or Component Names 4-Methylpyridin-2-amine
Shipping Condition Room Temperature
Molecular Formula C6 H8 N2
Certifications/Compliance ISO 17025
Synonym Rifaximin Imp. A (EP), 2-Pyridinamine, 4-methyl-, 4-Picoline, 2-amino- (7CI,8CI), (4-Methylpyridin-2-yl)amine, 2-Amino-4-methylpyridine, 2-Amino-4-picoline, 2-Amino-gamma-picoline, 4-Methyl-2-aminopyridine, 4-Methyl-2-pyridinamine, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine, Aminopicoline, Ascensil, NSC 1490, NSC 176165, NSC 6972, RA 1226, VMI 20-4, W 45, W 45 Raschig, Rifaximin Imp A (EP), Rifaximin Impurity A
Solution Type Neat
SMILES Cc1ccnc(N)c1
Molecular Weight (g/mol) 108.14
Quantity 4 x 25 mg
Formula Weight 108.144 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.