missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpiperidine, TRC

Product Code. 30464618
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464618 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464618 Supplier Toronto Research Chemicals Supplier No. TRCM32566550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylpiperidine
CAS 626-58-4
Molecular Formula C6H13N
Quantity 50 g
Synonym 4-Pipecoline, 4-Methylpiperidine, NSC 126910, NSC 66495, gamma-Pipecoline
InChI Formula InChI=1S/C6H13N/c1-6-2-4-7-5-3-6/h6-7H,2-5H2,1H3
SMILES CC1CCNCC1
IUPAC Name 4-methylpiperidine
Molecular Weight (g/mol) 99.17
Formula Weight 99.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.