missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphthalic anhydride, 96%

Product Code. 10587552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10587552 25 g Glass bottle 25g
10466062 5 g Glass bottle 5g
10509793 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10587552 Supplier Thermo Scientific Acros Supplier No. 414790250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19438-61-0 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-61-0
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00041856
InChI Key ZOXBWJMCXHTKNU-UHFFFAOYSA-N
Synonym 4-methylphthalic anhydride, 5-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-methyl, unii-2p2740fpck, phthalic anhydride, 4-methyl, dsstox_cid_7805, 5-methyl-1,3-dihydro-2-benzofuran-1,3-dione, dsstox_rid_78575, dsstox_gsid_27805, 4methylphthalic anhydride
PubChem CID 88069
IUPAC Name 5-methyl-2-benzofuran-1,3-dione
SMILES CC1=CC2=C(C=C1)C(=O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 88.0°C to 92.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Formula Weight 162.14
Percent Purity 96%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Methylphthalic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylphthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.