missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methylphenyl)methanesulfonyl chloride, Technical Grade, Maybridge
Specifications
Specifications
| CAS | 51419-59-1 |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C8H9ClO2S |
| Quantity | 1 g |
| Synonym | 4-methylbenzylsulfonyl chloride, 4-methylphenyl methanesulfonyl chloride, p-tolylmethanesulfonyl chloride, p-tolyl-methanesulfonyl chloride, p-tolyl methanesulfonyl chloride, 4-methylphenyl methyl sulfonyl chloride, pubchem5592, pubchem5631, acmc-209wgt, 4-methylbenzylsulfonylchloride |
| InChI Key | JALKUHLLMWYIAT-UHFFFAOYSA-N |
| SMILES | CC1=CC=C(C=C1)CS(=O)(=O)Cl |
| IUPAC Name | (4-methylphenyl)methanesulfonyl chloride |
| Molecular Weight (g/mol) | 204.668 |
| PubChem CID | 2794648 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?