missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals

Product Code. 15428139
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428139 1 g 1g
15438139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428139 Supplier Thermo Scientific Alfa Aesar Supplier No. J66686.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, warm methanol, DMSO, or DMF.

Notes
Store at 4°C. Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28244-98-6
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD04973631
InChI Key IQCLIQLFPVKINX-SJHCENCUSA-N
Synonym 4-methylphenylthio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyl oxane-3,4,5-triol, 4-methylphenyl b-d-thiogalactopyranoside, p-tolyl 1-thio-beta-d-galactopyranoside, 4-methylphenyl 1-thio-b-d-galactopyranoside, p-methylphenyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-methylphenyl sulfanyloxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triol
PubChem CID 7315241
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methylphenyl)sulfanyloxane-3,4,5-triol
SMILES CC1=CC=C(C=C1)SC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 137°C to 139°C
Color White
Quantity 1 g
Solubility Information Soluble in water,warm methanol,DMSO,or DMF.
Formula Weight 286.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methylphenyl beta-D-thiogalactopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.