missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (10 mg/mL in Methanol), TRC

Product Code. 30398772
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398772 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398772 Supplier Toronto Research Chemicals Supplier No. TRCKIT41081X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosonornicotine ketone
CAS 64091-91-4
Molecular Formula C10 H13 N3 O2
Quantity 1x1 mL
Synonym 1-Butanone, 4-(methylnitrosoamino)-1-(3-pyridinyl)-, 4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, 4-(Methyl-N-nitrosamino)-1-(3-pyridyl)-1-butanone, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone, 4-(Methylnitrosoamino)-1-(3-pyridyl)-1-butanone, 4-(N-Methyl-N-nitrosamino)-1-(3-pyridyl)-1-butanone, 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone, NNK, NNK (carcinogen), N-Nitrosonornicotine ketone
InChI Formula InChI=1S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8H,3,5,7H2,1H3
SMILES CN(CCCC(=O)c1cccnc1)N=O
IUPAC Name N-methyl-N-(4-oxo-4-pyridin-3-ylbutyl)nitrous amide
Molecular Weight (g/mol) 207.23
Formula Weight 207.1008
Concentration or Composition (by Analyte or Components) 10 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.