missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol (1 mg/mL in Methanol), TRC

Product Code. 30392671
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392671 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392671 Supplier Toronto Research Chemicals Supplier No. TRCKIT05651X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol
CAS 76014-81-8
Molecular Formula C10 H15 N3 O2
Quantity 1x1 mL
Synonym 3-Pyridinemethanol, α-[3-(methylnitrosoamino)propyl]-, α-[3-(Methylnitrosoamino)propyl]-3-pyridinemethanol, (±)-NNAL, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, 4-(Methylnitrosoamino)-1-(3-pyridyl)-1-butanol, 4-(N-Methyl-N-nitrosamino)-1-(3-pyridyl)butan-1-ol, NNAL, Racemic 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol, Racemic NNAL
InChI Formula InChI=1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3
SMILES CN(CCCC(O)c1cccnc1)N=O
IUPAC Name N-(4-hydroxy-4-pyridin-3-ylbutyl)-N-methylnitrous amide
Molecular Weight (g/mol) 209.24
Formula Weight 209.1164
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.