missing translation for 'onlineSavingsMsg'
Learn More

4-Methylnicotinic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 10500553
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10500553 10 g Amber Glass Bottle 10g
10101182 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10500553 Supplier Thermo Scientific Maybridge Supplier No. SB01942EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 94015-05-1
Molecular Formula C7H6NO2
Molecular Weight (g/mol) 136.13
MDL Number MFCD00832700
InChI Key ZKUZSTXNVMIDCY-UHFFFAOYSA-M
Synonym 4-methylnicotinic acid hydrochloride, 4-methylpyridine-3-carboxylic acid hydrochloride, 4-methylnicotinic acid hcl, 4-methyl-nicotinic acid hydrochloride, acmc-209wrr, 4-methyl nicotinic acid hydrochloride, 4-methylnicotinic acid hydrochloride;, 4-methylnicotinic acid monohydrochloride, 4-methylnicotinic acid hydrochloride salt, 4-methylnicotinic acid monohydrochloride;
PubChem CID 2796506
SMILES CC1=CC=NC=C1C([O-])=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 4-methylpyridine-3-carboxylate
Formula Weight 173.6
Percent Purity 95%
Chemical Name or Material 4-Methylnicotinic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.