missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-oxide, 50 wt.% solution in water

Product Code. 10735062 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 10735062

missing translation for 'mfr': Thermo Scientific Acros 348270010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylmorpholine N-oxide
Name Note 50 wt.% solution in water
CAS 7529-22-8, 7732-18-5
CAS Min % 50.0, 48.0
CAS Max % 52.0, 50.0
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 118.0°C to 119.0°C
Flash Point >100°C
Packaging Glass bottle
Molecular Formula C5H11NO2
MDL Number MFCD00005947
Quantity 1 kg
Fieser 07,244
Merck Index 15, 6171
Specific Gravity 1.13
Synonym 4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
Solubility Information Solubility in water: miscible
InChI Key LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES C[N+]1([O-])CCOCC1
IUPAC Name 4-methyl-4-oxidomorpholin-4-ium
Molecular Weight (g/mol) 117.15
PubChem CID 82029
ChEBI CHEBI:52093
Formula Weight 117.15
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylmorpholine N-oxide, 50% wt.% solution in water
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.