missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanone, TRC

Product Code. 30477732
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30477732 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30477732

Brand: Toronto Research Chemicals TRCM2948855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylcyclohexanone
CAS 589-92-4
Molecular Formula C7 H12 O
Quantity 5 g
Synonym 4-Methylcyclohexanone, 4-Methyl-1-cyclohexanone, NSC 3816
InChI Formula InChI=1S/C7H12O/c1-6-2-4-7(8)5-3-6/h6H,2-5H2,1H3
SMILES CC1CCC(=O)CC1
IUPAC Name 4-methylcyclohexan-1-one
Molecular Weight (g/mol) 112.17
Formula Weight 112.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.