missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanone, 98+%

Product Code. 11484584
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484584 25 g 25g
11494584 100 g 100g
11404594 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484584 Supplier Thermo Scientific Alfa Aesar Supplier No. A13186.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 589-92-4 | C7H12O | 112.17 g/mol

4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-92-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001643
InChI Key VGVHNLRUAMRIEW-UHFFFAOYSA-N
Synonym 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
PubChem CID 11525
IUPAC Name 4-methylcyclohexan-1-one
SMILES CC1CCC(=O)CC1

Specifications

Melting Point -41°C
Boiling Point 169°C to 171°C
Flash Point 48°C (118°F)
Quantity 25 g
UN Number UN2297
Beilstein 506746
Formula Weight 112.17
Percent Purity ≥98%
Chemical Name or Material 4-Methylcyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.