missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol, cis + trans, 98%

Product Code. 11492738
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492738 50 mL 50mL
11402748 250 mL 250mL
11482738 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492738 Supplier Thermo Scientific Alfa Aesar Supplier No. A16734.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-91-3 | C7H14O | 114.19 g/mol

2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001449,MFCD00064171,MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(O)CC1

Specifications

Melting Point °C to 41°C
Density 0.915
Boiling Point 171°C to 173°C
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 50 mL
Beilstein 1445998
Solubility Information Soluble in water (15mg/ml at 20°C),ether,alcohols,and many organic solvents.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 4-Methylcyclohexanol cis + trans, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.