missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylchrysoeriol, MedChemExpress

Product Code. 16477407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477407 5 mg 5mg
16455526 100 mg 100mg
16434698 10 mg 10mg
16420987 50 mg 50mg
16413206 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477407 Supplier Medchem Express Supplier No. HY112734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD.

4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD.

Specifications

Chemical Name or Material 4'-Methylchrysoeriol
CAS 4712-12-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H14O6
Quantity 5 mg
Solubility Information DMSO : 20.83 mg/mL (66.28 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 314.29
Formula Weight 314.29
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.