missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcatechol, 96%

Product Code. 11381607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381607 100 g 100g
11371607 25 g 25g
11391607 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381607 Supplier Thermo Scientific Alfa Aesar Supplier No. L04495.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 452-86-8 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Methylcatechol is used to synthesize antimicrobial, antioxidant. It also acts as effective inhibitor. It can be used to produce 4-bromo-5-methyl-pyrocatechol. It Increases Brain-Derived Neurotrophic Factor Content and mRNA Expression in Cultured Brain Cells.

Solubility
Soluble in water, methanol, acetone, ether, and alcohol.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-86-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002205
InChI Key ZBCATMYQYDCTIZ-UHFFFAOYSA-N
Synonym 4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
PubChem CID 9958
ChEBI CHEBI:17254
IUPAC Name 4-methylbenzene-1,2-diol
SMILES CC1=CC=C(O)C(O)=C1

Specifications

Melting Point 67°C to 69°C
Boiling Point 251°C
Flash Point 140°C (284°F)
Quantity 100 g
Beilstein 636512
Solubility Information Soluble in water,methanol,acetone,ether,and alcohol.
Formula Weight 124.14
Percent Purity 96%
Chemical Name or Material 4-Methylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.