missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-(Methylcarbamoyl)benzeneboronic acid, 98%
Chemical Identifiers
CAS | 121177-82-0 |
---|---|
Molecular Formula | C8H10BNO3 |
Molecular Weight (g/mol) | 178.982 |
MDL Number | MFCD03788427 |
InChI Key | UWKSYZHFTGONHY-UHFFFAOYSA-N |
Synonym | 4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836 |
PubChem CID | 2773526 |
IUPAC Name | [4-(methylcarbamoyl)phenyl]boronic acid |
SMILES | B(C1=CC=C(C=C1)C(=O)NC)(O)O |
Description
Chemical Identifiers
121177-82-0 | |
178.982 | |
UWKSYZHFTGONHY-UHFFFAOYSA-N | |
2773526 | |
B(C1=CC=C(C=C1)C(=O)NC)(O)O |
C8H10BNO3 | |
MFCD03788427 | |
4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836 | |
[4-(methylcarbamoyl)phenyl]boronic acid |
Specifications
121177-82-0 | |
C8H10BNO3 | |
4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836 | |
B(C1=CC=C(C=C1)C(=O)NC)(O)O | |
178.982 | |
178.98 | |
4-(Methylcarbamoyl)benzeneboronic acid |
166°C to 168°C | |
MFCD03788427 | |
UWKSYZHFTGONHY-UHFFFAOYSA-N | |
[4-(methylcarbamoyl)phenyl]boronic acid | |
2773526 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures