missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-carboxaldehyde, 96%

Product Code. 15447187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447187 250 mg 250mg
15437187 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447187 Supplier Thermo Scientific Alfa Aesar Supplier No. H34249.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36393-42-7 | C14H12O | 196.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36393-42-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD02258938
InChI Key BCINBWXQYBLSKO-UHFFFAOYSA-N
Synonym 4-4-methylphenyl benzaldehyde, 4'-methyl-1,1'-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carbaldehyde, 4'-methyl-4-formylbiphenyl, 1,1'-biphenyl-4-carboxaldehyde, 4'-methyl, 4'-methyl-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carboxaldehyde, 4'-methyl-biphenyl-4-carboxaldehyde, 4'-methyl 1,1'-biphenyl-4-carbaldehyde, pubchem10296
PubChem CID 1392762
IUPAC Name 4-(4-methylphenyl)benzaldehyde
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 108°C to 113°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 196.25
Percent Purity 96%
Chemical Name or Material 4'-Methylbiphenyl-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.