missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-4-carboxaldehyde, 96%

Product Code. 15437187
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15437187

Brand: Thermo Scientific Alfa Aesar H34249.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36393-42-7 | C14H12O | 196.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36393-42-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD02258938
InChI Key BCINBWXQYBLSKO-UHFFFAOYSA-N
Synonym 4-4-methylphenyl benzaldehyde, 4'-methyl-1,1'-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carbaldehyde, 4'-methyl-4-formylbiphenyl, 1,1'-biphenyl-4-carboxaldehyde, 4'-methyl, 4'-methyl-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carboxaldehyde, 4'-methyl-biphenyl-4-carboxaldehyde, 4'-methyl 1,1'-biphenyl-4-carbaldehyde, pubchem10296
PubChem CID 1392762
IUPAC Name 4-(4-methylphenyl)benzaldehyde
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 108°C to 113°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 196.25
Percent Purity 96%
Chemical Name or Material 4'-Methylbiphenyl-4-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.