missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylsulfonyl chloride, 97%

Product Code. 15254408 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254408 5 g 5g
15203558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254408 Supplier Thermo Scientific Acros Supplier No. 454840050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51419-59-1 | C8H9ClO2S | 204.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51419-59-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.68
InChI Key JALKUHLLMWYIAT-UHFFFAOYSA-N
Synonym 4-methylbenzylsulfonyl chloride, 4-methylphenyl methanesulfonyl chloride, p-tolylmethanesulfonyl chloride, p-tolyl-methanesulfonyl chloride, p-tolyl methanesulfonyl chloride, 4-methylphenyl methyl sulfonyl chloride, pubchem5592, pubchem5631, acmc-209wgt, 4-methylbenzylsulfonylchloride
PubChem CID 2794648
IUPAC Name (4-methylphenyl)methanesulfonyl chloride
SMILES CC1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Methylbenzylsulfonyl chloride
compliance-icons compliance-icons
Product Identifier
  • 4-Methylbenzylsulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.