missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine, 98%

Product Code. 11404824
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404824 25 g 25g
11414824 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404824 Supplier Thermo Scientific Alfa Aesar Supplier No. A13768.14
  • ONLINE EXCLUSIVE PRICE
    £42.60
    £38.65 / 25g
    Save £3.95
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-84-7 | C8H11N | 121.183 g/mol

The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-84-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008123
InChI Key HMTSWYPNXFHGEP-UHFFFAOYSA-N
Synonym 4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
PubChem CID 66035
IUPAC Name (4-methylphenyl)methanamine
SMILES CC1=CC=C(C=C1)CN

Specifications

Melting Point 12°C to 13°C
Density 0.955
Boiling Point 196°C to 197°C
Flash Point 75°C (167°F)
Refractive Index 1.535
Quantity 25 g
UN Number UN2735
Beilstein 956670
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 4-Methylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.