missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl isothiocyanate, 96%, Thermo Scientific Chemicals

Catalog Number 11387576
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
3694-46-0
C9H9NS
163.238
MFCD00041118
OAXIUBJXQISJEV-UHFFFAOYSA-N
4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference
19409
1-(isothiocyanatomethyl)-4-methylbenzene
CC1=CC=C(C=C1)CN=C=S
This item is not returnable. View return policy
3694-46-0
C9H9NS
163.238
MFCD00041118
OAXIUBJXQISJEV-UHFFFAOYSA-N
4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference
19409
1-(isothiocyanatomethyl)-4-methylbenzene
CC1=CC=C(C=C1)CN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.08
Boiling Point 97°C (1 mmHg)
Odor Pungent
Refractive Index 1.589
Quantity 2 g
UN Number UN2922
Beilstein 2082478
Sensitivity Moisture sensitive
Formula Weight 163.24
Percent Purity 96%
Chemical Name or Material 4-Methylbenzyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'