missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl cyanide, 98%

Product Code. 10447002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10447002 5 g Glass bottle 5g
10615562 25 g Glass bottle 25g
10590034 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10447002 Supplier Thermo Scientific Acros Supplier No. 126440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2947-61-7 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2947-61-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001922
InChI Key RNHKXHKUKJXLAU-UHFFFAOYSA-N
Synonym 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
PubChem CID 76280
IUPAC Name 2-(4-methylphenyl)acetonitrile
SMILES CC1=CC=C(CC#N)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 242°C to 243°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2CN
Refractive Index 1.5170 to 1.5195
Quantity 5 g
Beilstein 09, 530
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzyl cyanide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylbenzyl cyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.