missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzotrifluoride, 98%

Product Code. 10377271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10377271 5 g Glass bottle 5g
10255761 250 mg Glass bottle 250mg
10145800 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377271 Supplier Thermo Scientific Acros Supplier No. 308910050
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6140-17-6 | C8H7F3 | 160.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6140-17-6
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.14
MDL Number MFCD00075476
InChI Key LRLRAYMYEXQKID-UHFFFAOYSA-N
Synonym 4-methylbenzotrifluoride, 1-methyl-4-trifluoromethyl benzene, 4-trifluoromethyl toluene, p-methylbenzotrifluoride, alpha,alpha,alpha-trifluoro-p-xylene, benzene, 1-methyl-4-trifluoromethyl, 4-methyl-1-trifluoromethyl benzene, 1-methyl-4-trifluormethyl benzol, pubchem1659, p-trifluoromethyltoluene
PubChem CID 80230
IUPAC Name 1-methyl-4-(trifluoromethyl)benzene
SMILES CC1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1440g/mL
Boiling Point 129°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3C6H4CF3
Refractive Index 1.4250 to 1.427
Quantity 5 g
Specific Gravity 1.144
Formula Weight 160.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzotrifluoride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.