missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzhydrylamine hydrochloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-1.0 mmol/g on PS-DVB

Product Code. 11430770
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430770 5 g 5g
11440770 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430770 Supplier Thermo Scientific Alfa Aesar Supplier No. 044455.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5267-49-2 | C14H16ClN | 233.74 g/mol

Hydrochloride resins have been used to study the treatment of hyperphosphataemia in patients with end-stage renal failure. And also used for attachment of linkers in a similar manner to PL-AMS Resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5267-49-2
Molecular Formula C14H16ClN
Molecular Weight (g/mol) 233.74
MDL Number MFCD00801150
InChI Key NHTZSJKMWBONMD-UHFFFAOYNA-N
Synonym 4-methylphenyl phenyl methanamine hydrochloride, 4-methylphenyl phenyl methyl amine hydrochloride, 4-methylbenzhydrylamine hydrochloride, phenyl p-tolyl methanamine hydrochloride, 4-methylphenyl phenyl methyl amine hcl, 4-methylphenyl-phenylmethanamine hydrochloride
PubChem CID 2734294

Specifications

Color White to Yellow
Mesh Size 100 to 200 Mesh
Quantity 5 g
Solubility Information Suspended in DCM/DFM(1:1).
Physical Form Beads
Chemical Name or Material 4-Methylbenzhydrylamine hydrochloride, Polymer-supported, 1% cross-linked, 0.5 to 1.0 mmol/g on PS-DVB

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.