missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenethiol, TRC

Product Code. 30479972
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479972 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479972 Supplier Toronto Research Chemicals Supplier No. TRCM29051050G
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzenethiol
CAS 106-45-6
Molecular Formula C7H8S
Quantity 50 g
Synonym p-Toluenethiol, 1-Mercapto-4-methylbenzene, 4-Mercaptotoluene, 4-Methyl-1-thiophenol, 4-Methylphenyl Mercaptan, 4-Methylphenylthiol, 4-Methylthiophenol, 4-Thiocresol, 4-Toluenethiol, NSC 2227, NSC 229565, p-Mercaptotoluene, p-Methylbenzenethiol, p-Methylbenzenthiol, p-Methylphenyl Mercaptan, p-Methylphenylthiol, p-Methylthiophenol, p-Thiocresol, p-Thiolcresol, p-Tolyl mercaptan, p-Tolylthiol
InChI Formula InChI=1S/C7H8S/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3
SMILES Cc1ccc(S)cc1
IUPAC Name 4-methylbenzenethiol
Molecular Weight (g/mol) 124.2
Formula Weight 124.0347
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.