missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulphonylhydrazide, 97%

Product Code. 11405643
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11405643

Brand: Thermo Scientific Alfa Aesar A13529.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1576-35-8 | C7H10N2O2S | 186.23 g/mol

p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4,3-c]pyridines and 1,2,3-selenadiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-35-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00007588
InChI Key ICGLPKIVTVWCFT-UHFFFAOYSA-N
Synonym 4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide
PubChem CID 15303
IUPAC Name 4-methylbenzenesulfonohydrazide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN

Specifications

Melting Point 108°C to 111°C
Density 1.4
Quantity 100 g
UN Number UN3226
Beilstein 610130
Solubility Information Soluble in water,alcohol.
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl hydrazide
compliance-icons compliance-icons
Product Identifier
  • 4-Methylbenzenesulphonylhydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Self-reaC & Dtive substanC & De or mixture Type C & D
Hazard Statement
  • H242-Heating may cause a fire.
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : 4-Methylbenzenesulphonylhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.