missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulfonic Acid Monohydrate, TRC

Product Code. 30464696
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30464696 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30464696

Brand: Toronto Research Chemicals TRCM288740100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzenesulphonic Acid Hydrate
CAS 6192-52-5
Purity Grade Notes HPLC
Molecular Formula C7 H8 O3 S . H2 O
Quantity 100 g
Synonym 4-Methylbenzenesulphonic acid hydrate, Benzenesulfonic acid, 4-methyl-, hydrate (1:1), Benzenesulfonic acid, 4-methyl-, monohydrate (9CI), p-Toluenesulfonic acid, monohydrate (8CI), 4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, Tosic acid monohydrate, p-Toluenesulfonic acid monohydrate
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
SMILES O.Cc1ccc(cc1)S(=O)(=O)O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.22
Formula Weight 190.03
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.