missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzenesulfonic Acid Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methylbenzenesulphonic Acid Hydrate |
| CAS | 6192-52-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H8 O3 S . H2 O |
| Quantity | 25 g |
| Synonym | 4-Methylbenzenesulphonic acid hydrate, Benzenesulfonic acid, 4-methyl-, hydrate (1:1), Benzenesulfonic acid, 4-methyl-, monohydrate (9CI), p-Toluenesulfonic acid, monohydrate (8CI), 4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, Tosic acid monohydrate, p-Toluenesulfonic acid monohydrate |
| InChI Formula | InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2 |
| SMILES | O.Cc1ccc(cc1)S(=O)(=O)O |
| IUPAC Name | 4-methylbenzenesulfonic acid;hydrate |
| Molecular Weight (g/mol) | 190.22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?