missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic acid, 98%

Product Code. 11460973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460973 25 g 25g
11440973 1 g 1g
11450973 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460973 Supplier Thermo Scientific Alfa Aesar Supplier No. A13347.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5720-05-8 | C7H9BO2 | 135.96 g/mol

4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-05-8

Specifications

Melting Point 246°C to 254°C
Molecular Formula C7H9BO3
MDL Number MFCD00236047
Quantity 25 g
Beilstein 2935970
Synonym 2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
Solubility Information It is soluble in water.
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1B(O)O
IUPAC Name (2-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 151.96
PubChem CID 2733958
Formula Weight 135.96
Percent Purity 98%
Chemical Name or Material 4-Methylbenzeneboronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.