missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisole, 99%

Product Code. 10076283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076283 2.5 kg Glass bottle 2.5kg
10123940 5 g Glass bottle 5g
10337440 250 g Glass bottle 250g
10522381 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10076283 Supplier Thermo Scientific Acros Supplier No. 164660025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-93-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-93-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008413
InChI Key CHLICZRVGGXEOD-UHFFFAOYSA-N
Synonym 4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
PubChem CID 7731
IUPAC Name 1-methoxy-4-methylbenzene
SMILES CC1=CC=C(C=C1)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 174°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OCH3
Refractive Index 1.5100 to 1.513
Quantity 2.5 kg
Beilstein 06, 392
Specific Gravity 0.96
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alchol
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.