missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisole, 99%

Product Code. 11464633
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464633 500 mL 500mL
11454633 100 mL 100mL
11474633 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464633 Supplier Thermo Scientific Alfa Aesar Supplier No. A11572.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-93-8 | C8H10O | 122.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-93-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00008413
InChI Key CHLICZRVGGXEOD-UHFFFAOYSA-N
Synonym 4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
PubChem CID 7731
IUPAC Name 1-methoxy-4-methylbenzene
SMILES CC1=CC=C(C=C1)OC

Specifications

Melting Point -50°C
Density 0.968
Boiling Point 174°C to 175°C
Flash Point 58°C (136°F)
Odor Aromatic
Refractive Index 1.511
Quantity 500 mL
UN Number UN3271
Beilstein 1237336
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 4-Methylanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.