missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)pyridine, 99%

Product Code. 11395405
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11395405

Brand: Thermo Scientific Alfa Aesar L19626.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1121-58-0 | C6H8N2 | 108.14 g/mol

4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

Solubility
Soluble in water, acetone, ethanol, ether and benzene.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-58-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00152625
InChI Key LSCYTCMNCWMCQE-UHFFFAOYSA-N
Synonym 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
PubChem CID 123098
IUPAC Name N-methylpyridin-4-amine
SMILES CNC1=CC=NC=C1

Specifications

Melting Point 124°C to 128°C
Quantity 1 g
Beilstein 108213
Solubility Information Soluble in water,acetone,ethanol,ether and benzene.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 4-(Methylamino)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.